Learn More
Medchemexpress LLC Actinine chloride | 6249-56-5 | 99.9% | 181.66 | 10 MM 1 ML
SDP
Supplier:  Medchemexpress LLC HY113270A10MM/1ML
Encompass_Preferred
Actinine (γ-Butyrobetaine) chloride is an angiopathic substance produced as an intermediary metabolite by gut microbiota that feed on carnitine in dietary red meat. It is implicated in arteriosclerosis and long-term cardiovascular death.
  • Endogenous metabolite involved in carnitine metabolism.
  • Implicated in arteriosclerosis and long-term cardiovascular death.
  • Related to ketones, aldehydes, and acids.
  • Useful as a reference standard for natural products.
  • Relevant for cardiovascular, blood, cardio-cerebrovascular, metabolic, and endocrine disease research.
  • Can be used in studies involving metabolic enzymes/proteases.
Catalog No. 50-004-36213