Learn More
Cayman Chemical ActInomycIn D 50mg
SDP
Supplier:  Cayman Chemical 1142150 MG
Encompass_Preferred
A cyclic polypeptide-containing anticancer antibiotic that binds to DNA and blocks transcription by interfering with RNA synthesis; ~10 nM promotes cell death in a variety of tumors
Catalog No. 50-295-3699