Learn More
Medchemexpress LLC Actinomycin X2 (Actinomycin V) | 18865-48-0 | MFCD00076115 | 98.0% | 1269.40 | C62H84N12O17 | 1MG

Supplier: Medchemexpress LLC HY1257471MG
Actinomycin X2 is a cyclic peptide antibiotic used in research for antibacterial and anticancer studies, including apoptosis induction and mechanistic pathway analysis. It is supplied as an orange-red solid powder suitable for biochemical and cell-based assays.
- Cyclic peptide antibiotic with antibacterial and anticancer activity.
- Used in research for apoptosis induction and pathway analysis.
- Appearance: orange to red solid powder.
- Purity: 98.0%.
- Molecular weight: 1269.40 g/mol; formula C62H84N12O17.
- CAS number 18865-48-0.
- Storage: powder -80°C (2 years) or -20°C (1 year); in solvent -80°C (6 months) or -20°C (1 month).
- Pack size: 1 mg vial; additional sizes available by request.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.