Learn More
AdipoGen Actinonin (25 mg)
SDP
Supplier:  AdipoGen AGCN20161M025
Encompass_Preferred
Actinonin (25 mg), Adipogen Life Sciences. Chemical. CAS: 13434-13-4. Formula: C19H35N3O5. MW: 385.5. Synthetic. Antibiotic. Aminopeptidase M, N and leucine and alanine aminopeptidase inhibitor. Potent peptide deformylase (PDF) inhibitor. Inhibits MMP-1, MMP-2, MMP-3, MMP-7, MMP-8, MMP-9, MMP-10, MMP-12 and MMP-13, but not MMP-14. Anticancer compound. Antiproliferative. Shows antitumor activity in vitro and in vivo Apoptosis inducer. Meprin A inhibitor.
Catalog No. 50-168-7282