Learn More
Cayman Chemical ActIphenol 5mg
SDP
Supplier:  Cayman Chemical 331195 MG
Encompass_Preferred
A bacterial metabolite with antiviral activity; active against coxsackievirus B3 and influenza A (IC50s = 14.37 and 34.4 UG/ml, respectively); potentiates activity of miconazole against C. albicans in a disc assay at 50 UG/disc
Catalog No. 50-294-8068