Learn More
Cayman Chemical ActIvATd SureLIghtRPhycoe 5mg
SDP
Supplier:  Cayman Chemical 329115 MG
Encompass_Preferred
SMCC-Activated SureLight™ R-phycoerythrin Uses: Custom antibody labeling, fluorescence-activated cell sorting, flow cytometry, Luminex, and T-cell Labeling
Catalog No. 50-294-7217