Learn More
Medchemexpress LLC Activated EG3 Tail | 1380600-06-5 | C43H47N3O10 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY15340250MG
Encompass_Preferred
Activated EG3 Tail is used in the synthesis of exon jumping oligomer conjugates. It yields an oligomer conjugate that is complementary to a selected target site in the human dystrophin gene and can induce exon 51 skipping, making it suitable for muscular dystrophy research.
  • Used in synthesis of exon jumping oligomer conjugates
  • Yields an oligomer conjugate complementary to a selected target site in the human dystrophin gene
  • Induces exon 51 skipping
  • Applicable to the research of muscular dystrophy
Catalog No. 50-003-73374