Learn More
Medchemexpress LLC Activated T Subunit | 956139-30-3 | C31H34ClN4O5P | 250 MG
SDP
Supplier:  Medchemexpress LLC 250MGHY153406
Encompass_Preferred
Activated T Subunit can be used in the synthesis of exon jumping oligomer conjugates.
  • Oligomer conjugates complement selected target sites in the human anti-muscular atrophy protein gene and induce exon 51 jumping.
  • Can be used for research of muscular dystrophy.
Catalog No. 50-003-74290