Learn More
Medchemexpress LLC Activated T Subunit | 956139-30-3 | C31H34ClN4O5P | 50 MG

Supplier: Medchemexpress LLC HY15340650MG
Activated T Subunit is utilized in the synthesis of exon jumping oligomer conjugates. These conjugates complement specific target sites in the human anti-muscular atrophy protein gene, inducing exon 51 jumping. It is suitable for research related to muscular dystrophy, with a molecular weight of 609.05 and a purity of 99.64%. This product appears as a white to off-white solid.
- Utilized in the synthesis of exon jumping oligomer conjugates
- Induces exon 51 jumping
- Suitable for research related to muscular dystrophy
- Molecular weight of 609.05
- Purity of 99.64%
- White to off-white solid appearance
- For research use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.