Learn More
Seracare Life Sciences Inc ACTIVATOR SOLUTION
Supplier:  Seracare Life Sciences Inc 55700002
Encompass_Preferred
Activator Solution for Immunohistochemistry is an oxidizing reagent. It is a component of the HistoMark RED and HistoMark BLUE immunohistochemical staining systems for alkaline phosphatase.
Catalog No. 50-675-51