Learn More
Medchemexpress LLC Activin RIB/ALK-4 R 100ug | 100ug
SDP
Supplier:  Medchemexpress LLC HYP77294100UG
Encompass_Preferred
Activin RIB also known as activin receptor type-1B (ACVR1B) or ALK-4 is a type I transmembrane serine/threonine kinase receptor that is part of the TGF- receptor superfamily Activin binds to a type II activin receptor (ACVR2or ACVR2B) and then recruits ACVR1B[1][2][3] Activin RIB/ALK-4 Protein Rhesus Macaque (HEK293 Fc) is produced in HEK293 cells with a C-Terminal Fc-tag It consists of 100 amino acids (R27-E126)
Catalog No. 50-001-86384