Learn More
Genscript Corporation Activin RIIA/ACVR2A hFc Chimer
Supplier:  Genscript Corporation Z04389500
Encompass
Activin A receptor type 2A (ACVR2A) is a membrane receptor in the transforming growth factor- beta (TGF- signaling pathway which is involved in the regulation of cell proliferation migration and apoptosis Loss of ACVR2A has an important role in cancer progression and distant metastasis and may serve as a prognostic marker in patients with colon cancer
Catalog No. 50-000-58670