Learn More
Cayman Chemical ACtyl LysIn MoncLNal ntI 100ug
SDP
Supplier:  Cayman Chemical 32125100 UG
Encompass_Preferred
Immunogen: Acetyl lysine-BSA • Host: Rabbit • Species Reactivity: (+) Species-independent • Cross Reactivity: (+) Lysine-acetylated proteins; (-) Non-acetylated lysine residues, lysine residues with other modifications • Applications: ChIP, ICC, IHC, IP, WB
Catalog No. 50-294-7810