Learn More
Buehler AcuThin Blade, >HRC45, 12in [305mm]
Supplier:  Buehler 104361010
Encompass
The AcuThin series offers thin blades that have been developed to minimize the area of cutting thus reducing the amount of damage to the sample. These blades allow for precise, delicate abrasive sectioning with minimal surface damage.
Catalog No. NC3235622