Learn More
Medchemexpress LLC ACV Tripeptide | 32467-88-2 | 98.7% | 363.43 | 1 MG
SDP
Supplier:  Medchemexpress LLC HYP35191MG
Encompass_Preferred
ACV Tripeptide is a tripeptide and a dedicated intermediate in the biosynthetic pathway producing penicillin and cephalosporin classes of β-lactam natural products in bacteria and fungi. It is synthesized nonribosomally by the ACV synthetase (ACVS) enzyme and can be utilized for the biosynthesis of β-lactam antibiotics. This product contains the unusual amino acid, α-aminoadipic acid (α-AAA), as well as L-cysteine and D-valine.
  • Initial dedicated intermediate in β-lactam biosynthetic pathway
  • Synthesized nonribosomally by ACV synthetase (ACVS) enzyme
  • Can be utilized for biosynthesis of β-lactam antibiotics
  • Contains α-aminoadipic acid (α-AAA), L-cysteine, and D-valine
  • Appears as a white to off-white solid
  • For research use only
Catalog No. 50-004-00009