Learn More
Medchemexpress LLC ACV Tripeptide | 32467-88-2 | 98.7% | 363.43 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP35195MG
Encompass_Preferred
ACV Tripeptide is a tripeptide that serves as the initial dedicated intermediate in the biosynthetic pathway leading to the penicillin and cephalosporin classes of β-lactam natural products in bacteria and fungi. It is synthesized nonribosomally by the ACV synthetase (ACVS) enzyme. This product can be used for the biosynthesis of β-lactam antibiotics.
  • Initial dedicated intermediate in β-lactam biosynthesis
  • Synthesized nonribosomally by ACV synthetase
  • Contains unusual amino acid, α-aminoadipic acid (α-AAA), L-cysteine, and D-valine
  • Utilized for biosynthesis of β-lactam antibiotics
Catalog No. 50-003-97396