Learn More
Medchemexpress LLC HY-19327 100mg , ACY-738 CAS:1375465-91-0 Purity:98%
SDP
Supplier:  Medchemexpress LLC HY19327100MG
Encompass_Preferred
Medchemexpress, HY-19327 100mg ACY-738 CAS:1375465-91-0 Formula:C14H14N4O2 IC50: 1.7 nM (HDAC6), 94 nM (HDAC1), 128 nM (HDAC2), 218 nM (HDAC3) Purity:98% ACY-738 is a potent, selective and orally-bioavailable HDAC6 inhibitor, with an IC 50 s of 1.7 nM; ACY-738 also inhibits HDAC1, HDAC2, and HDAC3, with IC 50 s of 94, 128, and 218 nM. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3605
 Restricted Item