Learn More
Cayman Chemical AcyclovIr-d4 1mg
SDP
Supplier:  Cayman Chemical 262721 MG
Encompass_Preferred
An internal standard for the quantification of acyclovir by GC- or LC-MS
Catalog No. 50-293-5079