Learn More
CPC Scientific H-Val-Gln-Ala-Ala-Ile-Asp-Tyr-Ile-Asn-Gly-OH (ammonium salt) 25MG
SDP
Supplier:  CPC Scientific ACP001B
Encompass
A difficult peptide, synthesized for evaluating SPPS protocols.ONE-LETTER SEQUENCE: VQAAIDYINGMOLECULAR FORMULA: C47H74N12O16MOLECULAR WEIGHT:1063.18STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [66851-75-0], RESEARCH AREA: MiscellaneousREFERENCES: C. Arunan et al. Elsevier 56, 3005-3011 (2000)
Catalog No. 50-210-8589