Learn More
Medchemexpress LLC AD 01, a 24 amino acid peptide of FKBPL | 959961-23-0 | 100.0% | 2703.94 | 10 MG
SDP
Supplier:  Medchemexpress LLC HYP228410MG
Encompass_Preferred
AD 01 is a 24 amino acid peptide derived from FKBPL (FK506-binding protein like), known for its potent anti-angiogenic activity. It functions by binding to the CD44 receptor, thereby inhibiting tumor cell migration in a CD44-dependent manner. This compound has demonstrated significant anti-tumor effects in in vivo studies.
  • Possesses potent anti-angiogenic activity.
  • Binds to the CD44 receptor.
  • Inhibits tumor cell migration in a CD44-dependent manner.
  • Up-regulates CD44 expression in MDA-MB-231 and HMEC-1 cells (in vitro).
  • Exhibits anti-tumor activity in DU145 tumors in Balb-c severe compromised immuno-deficient (SCID) mice (in vivo).
Catalog No. 50-004-23015