Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC AD 01 | 959961-23-0 | 100.0% | 2703.94 | 5 MG
SDP

Supplier:  Medchemexpress LLC HYP22845MG

Encompass_Preferred

AD 01 is a 24 amino acid peptide of FKBPL (FK506-binding protein like) that possesses potent anti-angiogenic activity. It binds to the CD44 receptor and inhibits tumor cell migration in a CD44 dependent manner. This product is for research use only.

  • Purity: 99.98%
  • CAS number: 959961-23-0
  • Molecular weight: 2703.94
  • Molecular formula: C115H187N33O42
  • Appearance: Solid
  • Color: White to off-white
  • Sequence: Gln-Ile-Dir-Gln-Gln-Pro-Arg-Asp-Pro-Pro-Thr-Glu-Thr-Leu-Glu-Leu-Glu-Val-Ser-Pro-Asp-Pro-Ala-Ser
  • Sequence shortening: QIRQQPRDPPTETLELEVSPDPAS
  • Up-regulates CD44 expression in vitro
  • Exhibits anti-tumor activity in vivo
  • Solubility: ≥ 100 mg/mL (36.98 mM) in H2O

Catalog No. 50-004-23520


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.