Learn More
Molecular Devices Molecular Devices Absorbance Filter, 515 nm, 8 nm
Supplier: Molecular Devices 65900012
Absorbance bandpass filter suitable for microplate absorbance assays. Centered at 515 nm with an 8 nm bandwidth, this optical filter installs in filter slide slots and provides narrowband absorbance detection for plate-based assays. Compatible with FilterMax F3 and F5 multimode microplate readers.
- Center wavelength 515 nm.
- Bandwidth 8 nm.
- Filter type Absorbance bandpass.
- Application Microplate absorbance measurements.
- Form factor Slide-mounted filter for filter-based readers.
Description
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.