Learn More
Molecular Devices Molecular Devices Photometric Filter, 560 nm, Filter Slide Compatible
Supplier:  Molecular Devices 65900018
Encompass_Preferred
Replacement photometric filter for FilterMax F3 and F5 multi-mode microplate readers. Provides a fixed-wavelength photometric option centered at 560 nm for absorbance and ELISA assays. Sold as a single filter unit for installation in multi-position filter slides.
  • Fixed center wavelength of 560 nm.
  • Intended for photometric (absorbance) assays and ELISA.
  • Sold individually as a single filter unit.
  • Designed for installation in multi-position filter slides of microplate readers.
  • Used to restore or expand fixed-wavelength reading options.
Catalog No. 50-000-14469