Learn More
Vector Biolabs AD-CMV-GFP-U6-H-KAT5-SHRNA
Supplier:  Vector Biolabs 90001VITRO/Q015442
Encompass
Ad-CMV-GFP-U6-h-KAT5-shRNA (shADV-212630)Deliverable: in-vitro grade adenovirus* Buffer: DMEM w/2% glycerol* Volume: 2ml* Titer: 1-5e10 PFU/ml* Pre-validated Knockdown: Yes* Reporter: GFP
Catalog No. NC3940163