Learn More
Vector Biolabs AD-CMV-GFP-U6-SCRMB-SHRNA
Supplier:  Vector Biolabs 90500VITRO/Q015442
Encompass
Ad-CMV-GFP-U6-scrmb-shRNA* Deliverable: in-vitro grade adenovirus* Buffer: DMEM w/2% glycerol* Volume: 2ml* Titer: 1-5e10 PFU/ml* Reporter: GFP
Catalog No. NC3940167