Learn More
Chiral Technologies, Inc. CHIRALPAK® AD-H 30x250mm,5 micron. Compatible with HPLC (Normal Phase)
Supplier:  Chiral Technologies, Inc. 19375
Encompass
CHIRALPAK® AD-H 30x250mm,5 micron. Compatible with HPLC (Normal Phase)TECHNIQUES- Semi-Prep/Prep Columns - HPLC mode - Normal PhaseSELECTORS- Coated Polysaccharide phases
Catalog No. NC3236066
Add to Cart
Request a Quote