Learn More
Medchemexpress LLC Ad80 | 1384071-99-1 | 99.9% | 473.43 g·mol⁻¹ | C22H19F4N7O | 1 ML
SDP
Supplier:  Medchemexpress LLC HY10196310MM 1ML
Encompass_Preferred
AD80 is a multikinase inhibitor that potently inhibits RET, RAF, SRC, and S6K while exhibiting greatly reduced mTOR activity. It is used in research to probe kinase-driven signaling and to evaluate anti-proliferative effects in cancer cell models. The compound is supplied as a 10 mM solution in DMSO (1 mL) and is intended for in vitro research use.
  • Potent inhibition of RET, RAF, SRC, and S6K.
  • Reduced activity against mTOR compared with other multikinase inhibitors.
  • Suitable for in vitro kinase assays and cell-based signaling studies.
  • High purity suitable for biochemical and cellular experiments (99.9%).
  • Supplied as a ready-to-use 10 mM solution in DMSO, 1 mL.
Catalog No. 50-002-47969