Learn More
Research Products International Corp ADA 100 G

Supplier: Research Products International Corp A10030100.0
- Substance Name: [N-(Carbamoylmethyl) iminodiacetic acid)] [N-(2-Aceta-mido)iminodiacetic acid]
- CAS Number: 26239-55-4
- Molecular Formula: C6H10N2O5
- Molecular Weight: 190.2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.