Learn More
Medchemexpress LLC Adagrasib | 2326521-71-3 | MFCD32263433 | 99.6% | 604.12 | C32H35ClFN7O2 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1301495MG
Encompass_Preferred
Adagrasib is a mutation-selective, covalent KRAS G12C inhibitor used in preclinical research. It binds covalently to cysteine 12 of KRAS, locking the protein in an inactive GDP-bound conformation and inhibiting downstream signaling. Supplied as a high-purity solid for biochemical and cellular assays.
  • Mutation-selective KRAS G12C covalent inhibitor.
  • High purity suitable for analytical and biological studies.
  • Available in small milligram pack sizes for screening and dose-response studies.
  • Powder storage provides multi-year stability; solvent aliquots stable at ultra-low temperatures.
  • Molecular weight 604.12 and formula C32H35ClFN7O2 for reference.
Catalog No. 50-002-62095