Learn More
Medchemexpress LLC Adamantan-1-ol | 768-95-6 | MFCD00074729 | 99.6% | 152.23 g·mol⁻¹ | C10H16O | 5g
SDP
Supplier:  Medchemexpress LLC HYY05315G
Encompass_Preferred
1-Adamantanol is a cyclic molecule with a hydroxyl group that can be used as an intermediate in the synthesis of adamantane 1-Adamantanol can be oxidized to 1 3-adamantanediol by the Streptomyces SA8 oxidation system[1]
Catalog No. 50-002-69059