Learn More
Medchemexpress LLC Adamantane-1,3,5-triol | 99181-50-7 | MFCD08276262 | 97.8% | 184.23 g/mol | C10H16O3 | 10 G
SDP
Supplier:  Medchemexpress LLC HYW01628710G
Encompass_Preferred
Adamantane-1,3,5-triol is a polycyclic triol used as a chemical intermediate for the synthesis of bioactive small molecules. The compound combines a rigid adamantane core with three hydroxyl functional groups, enabling selective derivatization for medicinal chemistry research and specialty materials applications.
  • Polycyclic cage scaffold provides rigidity and lipophilicity
  • Three hydroxyl groups enable esterification, etherification, and other derivatizations
  • Suitable as an intermediate in small-molecule synthesis and medicinal chemistry
  • Supplied with batch-specific purity (HPLC 97.8%) and certificate of analysis
  • Available in small laboratory quantities for research and development
Catalog No. 50-003-02672