Learn More
Medchemexpress LLC Adamantane | 281-23-2 | 50 G
Supplier: Medchemexpress LLC HYN242750G
Adamantane is a polycyclic cage molecule with high symmetry and remarkable properties. It can be incorporated into a lipophilic part of the lipid bilayer that constitutes membranes and as an anchor in the lipid bilayer of liposomes. Adamantane can be studied in pharmacological research to improve the lipophilicity and pharmacological properties of other active products. It can also be used as a skeleton to synthesize compounds with antibacterial effects.
- Polycyclic cage molecule with high symmetry and remarkable properties
- Can be incorporated into a lipophilic part of the lipid bilayer
- Can be used as an anchor in the lipid bilayer of liposomes
- Studied in pharmacological research to improve lipophilicity and pharmacological properties
- Used as a skeleton to synthesize compounds with antibacterial effects