Learn More
Medchemexpress LLC Adamantyl urea | 1338780-86-1 | 98.2% | 324.34 g/mol | C17H19F3N2O | 25 MG

Supplier: Medchemexpress LLC HY10186725MG
AU1235 is an adamantyl urea that potently inhibits MmpL3, a membrane transporter essential for Mycobacterium tuberculosis cell wall biosynthesis. It exhibits activity against M. tuberculosis, including multidrug-resistant isolates, and is supplied for research use with specified purity, solubility, and storage recommendations.
- Purity 98.2%.
- CAS number 1338780-86-1.
- Molecular weight 324.34 g/mol; formula C17H19F3N2O.
- Appearance solid, white to off-white.
- Solubility soluble in DMSO at 25 mg/mL with sonication.
- Storage powder: -20°C (3 years) or 4°C (2 years); in solvent: -80°C (2 years) or -20°C (1 year).
- Activity active against Mycobacterium tuberculosis, including multidrug-resistant isolates.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.