Learn More
Medchemexpress LLC ADAMTS-5 Inhibitor 5mg | 929634-33-3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1149965MG
Encompass_Preferred
ADAMTS-5 inhibitor is a small-molecule research compound that selectively inhibits ADAMTS-5 (aggrecanase-2) with an IC50 of 1.1 μM. It is supplied as a high-purity solid for biochemical and cell-based studies.
  • Potent ADAMTS-5 inhibition (IC50 1.1 μM).
  • Greater than 40-fold functional selectivity over ADAMTS-4.
  • High reported purity (99.59%).
  • Molecular weight 449.92 g/mol; formula C16H11ClF3N3OS3.
  • Available in mg-scale quantities for research use.
Catalog No. 50-003-08075