Learn More
Cayman Chemical ADAMTS-5 InhIbItr 500ug
SDP
Supplier:  Cayman Chemical 17188500 UG
Encompass_Preferred
An inhibitor of ADAMTS-5 (aggrecanase-2; IC50 = 1.1 µM) with 40-fold selectivity over ADAMTS-4 (aggrecanase-1)
Catalog No. 50-296-6729