Learn More
Aviva Systems Biology Corp ADAMTS19 Antibody - N-terminal region (ARP39809_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP39809_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ADAMTS19. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human ADAMTS19. Alternate names: FLJ16042ApplicationWB. Species reactivity: Cow, Dog, Human, Rat
Catalog No. 50-284-831