Learn More
Ace Glass, Inc. Column end fitting, #15 to 1/4-28 UNF helicoil tap, porosity D (10-20 micron) glass filter disc included
SDP
Supplier:  Ace Glass, Inc. 580157
Encompass_Preferred
#15 Ace-Thred column end fitting adapter with -28UNF tap with 18-8 stainless helicoil insert for durability. Design allows for a leak-tight seal without the need for an O-ring. Porosity D (10-20 micron) glass filter disc included.
Catalog No. 50-978-438