Learn More
Medchemexpress LLC Adapalene | 106685-40-9 | 99.9% | C28H28O3 | 100 MG
SDP
Supplier:  Medchemexpress LLC HYB0091100MG
Encompass_Preferred
Adapalene is a third-generation synthetic retinoid widely used for acne research. It acts as a potent RAR agonist, with AC50s of 2.3 nM for RARβ, 9.3 nM for RARγ, and 22 nM for RARα. This compound also non-competitively inhibits the enzymatic activity of GOT1 and exhibits anti-tumor activity.
  • Third-generation synthetic retinoid
  • Potent RAR agonist for RARβ, RARγ, and RARα
  • Non-competitive inhibitor of GOT1 enzymatic activity
  • Exhibits anti-tumor activity
  • Induces apoptosis and inhibits proliferation in ES-2 cells
  • Inhibits growth of DLD1 cell-derived xenograft tumors
Catalog No. 50-004-32190