Learn More
Medchemexpress LLC Adapalene | 106685-40-9 | 99.94% | C28H28O3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB009110MM/1ML
Encompass_Preferred
Adapalene (CD271) is a third-generation synthetic retinoid primarily used for acne research. It acts as a potent RAR agonist and non-competitively inhibits the enzymatic activity of GOT1, also exhibiting anti-tumor activity. It is supplied as a 10 mM solution in DMSO, ready for reconstitution.
  • Potent RAR agonist
  • Inhibits GOT1 enzymatic activity
  • Exhibits anti-tumor activity
  • Supplied as a 10 mM solution in DMSO
Catalog No. 50-004-32551