Learn More
Beckman Coulter Adapter, 870329
Supplier:  Beckman Coulter 870329
Encompass_Preferred
The Beckman Coulter Microfuge 18 Centrifuge Tube Adapter, catalog number 870329, is designed to adapt 0.5mL and 1.5mL microcentrifuge tubes to the Microfuge 18 Centrifuge rotor. It allows centrifugation of up to 6 microcentrifuge tubes at a time. The adapter is made of durable polypropylene and can withstand centrifugal forces up to 18,000 x g. It fits securely into the Microfuge 18 rotor and works with most major brands of microcentrifuge tubes. This adapter provides a convenient way to spin down small volume samples in a standard lab microcentrifuge.
Catalog No. NC2067538