Learn More
Chemglass Life Sciences Adapter, Thermometer, Inlet, 45/50 Inner Joint, 95mm Height, #15 Chem-Thread
SDP
Supplier:  Chemglass Life Sciences CG104246
Encompass_Preferred
Adapter, Thermometer, Inlet, 45/50 Inner Joint, 95mm Height, #15 Chem-Thread. Adapter has a #7 or #15 Chem-Thread at the top for vacuum tight seal of plain stem thermo-meters or any other tube having an O.D. between 4 and 7mm. Supplied complete with a Celcon compression cap and Viton o-ring.
Catalog No. CG104246