Learn More
Chemglass Life Sciences 160° Adapter, 65/40 Socket Joint to 80 mm Flange - for CG-1968-D distillation kits
SDP
Supplier:  Chemglass Life Sciences CG1968D300
Encompass_Preferred
The Chemglass 160° adapter converts an 80 mm flat flange to a 65/40 polished socket joint at a 160° orientation. It is designed for use in distillation and reactor assemblies, including diagonal distillation cart systems, to provide a secure glass-to-glass connection between flanged vessels and tapered joints.
  • 160° angle for off-axis connections.
  • Converts an 80 mm flat flange to a 65/40 socket joint.
  • Polished finish for smooth glass-to-glass sealing surfaces.
  • Designed for use in distillation and reactor assemblies.
Catalog No. 50-273-3554