Learn More
World Precision Instrument Adapter Cable
SDP
Supplier:  World Precision Instrument 3578
Encompass
Cable for Electrophysiology
This cable has a connector for use with Ag/AgCl (silver/silver chloride) pellts and an other connector for pellets.
Catalog No. 50-822-067