Learn More
Chemglass Life Sciences Adapter, Claisen, 14/20 Joint, Modified
SDP
Supplier:  Chemglass Life Sciences CG107414
Encompass_Preferred
Adapter, Claisen, 14/20 Joint, Modified. Claisen style adapter with an additional standard taper outer joint at a 45º angle to the vertical outer joint.
Catalog No. 50-872-917