Learn More
Chemglass Life Sciences 14/20 Adapter, Connecting, Height 50mm
SDP
Supplier:  Chemglass Life Sciences CG1055A14
Encompass_Preferred
14/20 Adapter, Connecting, Height 50mm. Adapter has a luer outer on top and a lower standard taper inner joint.
Catalog No. 50-153-0998