Learn More
Chemglass Life Sciences Adapter, Connecting, with Frit, Airfree, Schlenk, 24/40 Joint Size
SDP
Supplier:  Chemglass Life Sciences AF0070F02
Encompass_Preferred
Adapter, Connecting, with Frit, Airfree, Schlenk, 24/40 Joint Size. Adapter has a coarse frit sealed between the #15 o-ring joint and the standard taper inner joint. Frit permits filtration of large particles before introduction into the flask and also serves as a bump trap for reactions being performed in flasks while connected to the manifold. Each adapter is supplied complete with one #116 Viton o-ring.
Catalog No. AF0070F02