Learn More
Chemglass Life Sciences Adapter, Distillation, 75°, 14/20 Inner Joints, Tooled Rubber Fitting
SDP
Supplier:  Chemglass Life Sciences CG102614
Encompass_Preferred
Adapter, Distillation, 75, 14/20 Inner Joints, Tooled Rubber Fitting. Adapter has a tubulation at the top for use with a CG-1040 rubber fitting. Fitting permits insertion of plain stem, partial immersion thermometers. Supplied complete with one CG-1040 rubber fitting. With the same size inner joints on the bottom and sidearm at a 75 angle to one another. FOR THERMOMETERS SEE CG-3503.
Catalog No. 50-153-0988