Learn More
World Precision Instrument Adapter Electrode 0.031 for DAM80P
SDP
Supplier:  World Precision Instrument 5469
Encompass
  • Adapter for the DAM80P probe
  • Mini banana plug (on the DAM80) to 0.79 mm (0.031") socket (for the metal microelectrode)
Catalog No. 50-252-1249