Learn More
Chemglass Life Sciences Adapter, Connecting, 29/42 Top Outer, 24/40 Lower Inner, Enlarging
SDP
Supplier:  Chemglass Life Sciences CG100207
Encompass_Preferred
Adapter, Connecting, 29/42 Top Outer, 24/40 Lower Inner, Enlarging. Used to connect dissimilar standard taper joint sizes. Lower inner joint is smaller in size than the top outer joint.
Catalog No. CG100207