Learn More
Cole-Parmer Dolan Jenner 020863050003 Light Pipe Adapter for High Output Fiber Optic Illuminator
Supplier:  Cole-Parmer 0974140
Encompass_Preferred
Parts and accessories for your Dolan Jenner lamps and fiber optic illuminators. Required for use with illuminators 41724-00 and 41724-05, and light pipes 09742-60 and 09742-70.
Catalog No. 50-628-563